missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gardner Denver Welch™ Vacuum Regulator with Dial Gauge for MP/MPC pumps
Mounts to the inlet of the MP/MPC pump to regulate the vacuum level by way of bleed valve.
Brand: Gardner Denver Welch™ 700458-02
This item is not returnable.
View return policy
Description
- For use with MP/MPC 095 Z, 110 E, MPC 105 T, MPC 155 Z pumps
Specifications
Vacuum Regulator with Dial Gauge | |
MP/MPC 095 Z, 110 E, MPC 105 T, MPC 155 Z pumps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gardner Denver Welch™ Vacuum Regulator with Dial Gauge for MP/MPC pumps >
Spot an opportunity for improvement?Share a Content Correction