Learn More
BUCHI Vacuum Seal
Use in conjunction with Buchi seal holder to create a vacuum seal in Buchi Rotavapors. BUCHI Vacuum Seals are designed for use with R-176/186/187/220/250 Rotavapors.
Brand: BUCHI 041095
Code : 62
Additional Details : Weight : 0.06000kg
Description
- For use with BUCHI Rotary evaporators
Specifications
Vacuum Seal | |
For R-176/186/187/220/250 |