missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valaciclovir hydrochloride hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 25 g |
Formula Weight | 360.8 |
MDL Number | MFCD01861507 |
InChI Key | ZCDDBUOENGJMLV-VGVXOIKUNA-N |
SMILES | Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O |
IUPAC Name | 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride |
Molecular Weight (g/mol) | 360.80 |
Chemical Name or Material | Valaciclovir hydrochloride hydrate |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'