Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Brand: Thermo Scientific™ U-400X
Code : 16
Additional Details : Weight : 0.05000kg
Description
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
- Designed for 10-32 port configurations
- Burr and contaminant free
Specifications
HPLC Injectors with 10-32 Port Configuration | |
316 Stainless Steel Male Nut for 1/16” tubing (0.0625in) | |
High Pressure Male Hex Nut | |
10 Pack |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.