missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Valeric Acid, 99%

Product Code. 10686394 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
5mL
This item is not returnable. View return policy

Product Code. 10686394

Brand: Thermo Scientific Chemicals 149570025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-52-4 | C5H10O2 | 102.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-52-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004413
InChI Key NQPDZGIKBAWPEJ-UHFFFAOYSA-N
Synonym valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n
PubChem CID 7991
ChEBI CHEBI:17418
IUPAC Name pentanoic acid
SMILES CCCCC(=O)O

Specifications

Melting Point -20.0°C to +18.0°C
Boiling Point 185.0°C
Color Colorless to Yellow
Assay Percent Range 99%
Linear Formula CH3(CH2)3CO2H
Beilstein 02, 299
Merck Index 15, 10090
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 102.13
Percent Purity 99%
Density 0.9400g/mL
Quantity 2.5 L
Flash Point 86°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4070 to 1.4100
Specific Gravity 0.94
Chemical Name or Material Valeric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.