Learn More
Ohaus™ Valor™ 7000 Compact Food Scale
The best compact food scale in its class. Ohaus™ Valor™ 7000 Compact Food Scale features dual displays and touchless sensors, a user-friendly menu and setup, and fast and accurate results to provide for a seamless and hassle-free user experience that is second to none.
Brand: Ohaus™ 30031822
508.50 GBP valid until 2024-08-31
Use promo code "21622" to get your promotional price.
Code : NEW
Additional Details : Weight : 4.90500kg Transport : UN number : 2800 Chem class : 8
Description
The Ohaus Valor 7000 compact food scale is the newest scale in Ohaus’s line of reliable and durable products that will increase food weighing and proportioning productivity. The advanced, user-friendly features provide for a seamless user experience.
- Front and rear touchless sensors control operation of certain functions to minimize food cross-contamination
- Food-safe design is NSF Certified, USDA-AMS Accepted, and supports HACCP-Certified systems
- Large front and rear backlit displays enhance the user experience and allow two operators to share one scale
- Rapid, one second stabilization time produces precise results, while also greatly increasing throughput and eliminating wait time
- Stainless steel weighing platform and sturdy ABS housing can stand up to daily use under the most demanding conditions
- Large, non-slip leveling feet grip the counter and prevent the scale from sliding during use
- Flexible connectivity: Standard RS232 with optional Ethernet, second RS232 or USB offers data transfer capabilities to multiple peripherals
- Integrated carrying handle and rechargeable lead acid battery enables uninterrupted operation wherever it is needed
Specifications
Built-in RS232, optional 2nd RS232, USB, Ethernet | |
Rechargeable (included) | |
NSF, USDA-AMS, HACCP, NTEP | |
ABS housing, stainless steel platform, touchless sensor, transportation handle, menu lock switch, up front level indicator | |
Dual LCD with White LED Backlight + 3 LED | |
Mechanical | |
V71P1502T | |
0.100000 g | |
1 sec. | |
-10°C to +40°C | |
kg, g, lb, oz, lb:oz | |
311 × 327 × 117 mm | |
311.00 mm | |
12.90 in. | |
300 mm | |
117.000 mm | |
311.00 mm | |
100/240 VAC 50/60 Hz internal universal power supply, internal rechargeable lead acid battery | |
50/60 Hz |
210 Hours with Rechargeable Battery | |
External | |
Ethernet (Available as an Accessory)USB (Available as an Accessory)RS232 (Included)2nd RS232 (Available as an Accessory) | |
1.5 kg | |
0.050000 g | |
0.100000 g | |
IP43 | |
Front and rear | |
14°F to 104°F | |
Valor™ 7000 Compact Food Scale | |
311.00 x 327.00 mm | |
12.20 in. | |
225 mm | |
327.00 mm | |
4.60 in. | |
12.20 in. | |
4.2 kg | |
100/240 V |