missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Valve Lockout Kit
Valve Lockout Kit for taps and electrical hazards.
£640.00 - £655.00
Specifications
Product Type | Valve Lockout Kit |
---|---|
For Use With (Application) | Lockout Tagout |
Quantity | 1 Ea. |
Product Code | Brand | Displayed Text | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Includes | Price | Quantity & Availability | ||||
11766795
|
Brady™
806187 |
Do not switch on | 6 Red Safety Padlocks, 12 Red Safety Lockout Hasps, Lockout Satchel, Pen, 10 Lockout Labels - Do not switch on |
£655.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
12708029
|
Brady™
806175 |
Do not operate | 1 Pack of Red Safety Padlocks, 1 Pack of Red Safety Lockouts, Do Not Operate Tag |
£640.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
Description
- Includes safety padlocks and lockout tags.
Specifications
Valve Lockout Kit | |
Lockout Tagout | |
1 Ea. |