Learn More
VAMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15546-1-AP-150UL
 View more versions of this product
Product Code. 16884764
  • £349.00 / 150µL
Explore more special offers
Description
Description
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
Specifications
Specifications
VAMP8
Polyclonal
Unconjugated
VAMP8
EDB, Endobrevin, VAMP 8, VAMP8
Rabbit
Antigen Affinity Chromatography
RUO
22320, 83730, 8673
-20°C
Liquid
Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70404, Q9BV40, Q9WUF4
VAMP8
VAMP8 Fusion Protein Ag7903
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote