missing translation for 'onlineSavingsMsg'
Learn More

Vanadium carbide, 99% (metals basis)

Product Code. 11366508
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11366508

Brand: Thermo Scientific Alfa Aesar 012141.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12070-10-9 | CV | 62.95 g/mol

Vanadium carbide is used as an additive to tungsten carbide to fine the carbide crystals to improve the property of the cermets. It used in cutting tools and the steel industry. Used as material for research in electronics. Used anode materials for lithium ion batteries, and in supercapacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-10-9
Molecular Formula CV
Molecular Weight (g/mol) 62.95
MDL Number MFCD00049698
InChI Key ZLANVVMKMCTKMT-UHFFFAOYSA-N
IUPAC Name methanidylidynevanadiumylium
SMILES [C-]#[V+]

Specifications

Melting Point 2810°C
Color Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% metal basis
Solubility Information Insoluble in water.
Formula Weight 62.95
Odor Odorless
Chemical Name or Material Vanadium carbide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.