Learn More
Vanadium(III) chloride, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 250.0°C |
Color | Black to Blue |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 96% min. (Argentometry) |
Linear Formula | VCl3 |
Fieser | 13,289 |
Solubility Information | Solubility in water: vigorous reaction. Other solubilities: soluble in abs.alcohol and ether |
Formula Weight | 157.3 |
Percent Purity | 97% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0449242 | Certificate of Analysis | 11/07/2024 | 197001000,197000250,197000050 |
A0463529 | Certificate of Analysis | 07/06/2024 | 197001000,197000250,197000050 |
A0449242 | Certificate of Analysis | 11/01/2023 | |
A0384528 | Certificate of Analysis | 15/12/2022 | 197001000,197000250,197000050 |
A0402141 | Certificate of Analysis | 14/12/2022 | 197001000,197000250,197000050 |
Safety and Handling


- Vanadium(III) chloride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
15Minutes
42Seconds
57
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.