missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) chloride, 97%

Product Code. 10791851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10791851

Brand: Thermo Scientific Acros 197001000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vanadium(III) chloride, 97%, Cl3V, CAS Number-7718-98-1, Vanadium trichloride, 100g, 96% min. (Argentometry), Black to Blue, 3.0000g/mL, 231-744-6, 13,289, 157.3, GHS Signal Word: Danger | CAS: 7718-98-1 | Cl3V | 157.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-98-1
Molecular Formula Cl3V
Molecular Weight (g/mol) 157.29
MDL Number MFCD00011454
InChI Key HQYCOEXWFMFWLR-UHFFFAOYSA-K
Synonym Vanadium trichloride
PubChem CID 62647
IUPAC Name trichlorovanadium
SMILES [Cl-].[Cl-].[Cl-].[V+3]

Specifications

Melting Point 250.0°C
Color Black to Blue
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min. (Argentometry)
Linear Formula VCl3
Fieser 13,289
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in abs.alcohol and ether
Formula Weight 157.3
Percent Purity 97%
Packaging Glass bottle
Specific Gravity 3
Density 3.0000g/mL
Chemical Name or Material Vanadium(III) chloride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vanadium(III) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.