missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) fluoride, 98%

Product Code. 11324919
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11324919

Brand: Thermo Scientific Alfa Aesar 081115.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-12-4 | F3V | 107.94 g/mol

Employed in oxygen-sensitive applications, such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-12-4
Molecular Formula F3V
Molecular Weight (g/mol) 107.94
MDL Number MFCD00042553
InChI Key PASVSMBJEYQYSN-UHFFFAOYSA-K
Synonym Vanadium trifluoride
IUPAC Name vanadium(3+) trifluoride
SMILES [F-].[F-].[F-].[V+3]

Specifications

Melting Point ≥1406°C
Physical Form Powder
Quantity 10 g
UN Number UN2923
Merck Index 14,9922
Solubility Information Practically insoluble in water, acetone, and glacial acetic acid
Formula Weight 107.94
Percent Purity 98%
Sensitivity Moisture sensitive
Density 3.363 g/mL at 19°C
Chemical Name or Material Vanadium (III) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.