Learn More
Vanadium(III) oxide, 95%, Thermo Scientific Chemicals
£95.27 - £327.63
Chemical Identifiers
CAS | 1314-34-7 |
---|---|
Molecular Formula | O3V2 |
Molecular Weight (g/mol) | 149.88 |
MDL Number | MFCD00011455 |
InChI Key | KFAFTZQGYMGWLU-UHFFFAOYSA-N |
Synonym | vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basisShow More |
PubChem CID | 518710 |
SMILES | O=[V]O[V]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351659
|
Thermo Scientific Alfa Aesar
012301.30 |
250 g |
£95.27
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341659
|
Thermo Scientific Alfa Aesar
012301.A1 |
1 kg |
£327.63
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-34-7 | |
149.88 | |
KFAFTZQGYMGWLU-UHFFFAOYSA-N | |
518710 |
O3V2 | |
MFCD00011455 | |
vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basisShow More | |
O=[V]O[V]=O |
Specifications
1970°C | |
Black | |
95% | |
MFCD00011455 | |
Insoluble in water | |
O=[V]O[V]=O | |
149.88 | |
149.88 | |
4.87 |
1314-34-7 | |
Powder | |
O3V2 | |
vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis | |
KFAFTZQGYMGWLU-UHFFFAOYSA-N | |
oxo[(oxovanadio)oxy]vanadium | |
518710 | |
Odorless | |
Vanadium(III) oxide |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P321-P330-P332+P313-P361-P363-P501c
H302-H311-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-230-9
missing translation for 'rtecsNumber' : YW3050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Vanadium(III) oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Vanadium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.