missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) oxide, 99.7% (metals basis)

Product Code. p-4408205
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11355647

Brand: Thermo Scientific Alfa Aesar 043377.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-34-7 | O3V2 | 149.88 g/mol

Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-34-7
Molecular Formula O3V2
Molecular Weight (g/mol) 149.88
MDL Number MFCD00011455
InChI Key KFAFTZQGYMGWLU-UHFFFAOYSA-N
Synonym vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis
PubChem CID 518710
IUPAC Name oxo(oxovanadiooxy)vanadium
SMILES O=[V]O[V]=O

Specifications

Melting Point 1970°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.7% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 149.88
Density 4.87
Chemical Name or Material Vanadium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.