missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(IV) etioporphyrin III oxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
Description
Vanadium(IV) etioporphyrin III oxide is used to check the trapping efficiency of the external AlOxlayer in AlOxcoating of ultrastable Zeolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 15709-03-2 |
Color | Purple |
Physical Form | Crystalline |
Quantity | 50 mg |
Molecular Formula | C32H38Cl2N4OV |
MDL Number | MFCD00058901,MFCD00058901 |
Synonym | oxovanadium 2+ ;2,7,12,17-tetraethyl-3,8,13,18-tetramethylporphyrin-22,24-diide |
Solubility Information | Insoluble in water. |
InChI Key | LLVNESQRTPCYSP-UHFFFAOYSA-N |
SMILES | Cl.Cl.O=[V].CCC1=C(C)C2=CC3=NC(=CC4=NC(=CC5=C(CC)C(C)=C([N-]5)C=C1[N-]2)C(C)=C4CC)C(C)=C3CC |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vanadium(IV) etioporphyrin III oxide, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction