missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) fluoride, 95%

Product Code. p-7048813
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11327247

Brand: Thermo Scientific Alfa Aesar 011543.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10049-16-8

Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-16-8
MDL Number MFCD00042554
Synonym vanadium iv fluoride, acmc-20aja2, vanadium 4+ tetrafluoride, vanadium 4+ ion tetrafluoride, fluoride, fluoride, fluoride, vanadium salt, fluoride

Specifications

Melting Point 325°C (decomposition)
Color Yellow to Brown
Physical Form Powder
Quantity 5 g
Linear Formula VF4
UN Number UN2923
Solubility Information Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
Formula Weight 126.94
Percent Purity 95%
Sensitivity Moisture sensitive
Density 2.975 g/mL
Chemical Name or Material Vanadium(IV) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.