missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) oxide meso-tetraphenylporphine, Thermo Scientific Chemicals

Product Code. 11314497
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11314497

Brand: Thermo Scientific Alfa Aesar 039583.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used for research in catalysis. Thin films of VOTPP may be used in detecting methane gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14705-63-6
Molecular Formula C44H30N4O2V
Molecular Weight (g/mol) 697.692
MDL Number MFCD00012154
InChI Key LRQXAIDMLNPKOR-UHFFFAOYSA-N
Synonym 5,10,15,20-tetraphenyl-21h,23h-porphine vanadium iv oxide
PubChem CID 131699123
IUPAC Name oxygen(2-);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;vanadium(4+)
SMILES C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[O-2].[O-2].[V+4]

Specifications

Color Purple
Physical Form Crystalline
Quantity 500 mg
Solubility Information Insoluble in water.
Formula Weight 679.67
Odor Odorless
Chemical Name or Material Vanadium(IV) oxide meso-tetraphenylporphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.