Learn More
Vanadium(IV) oxide phthalocyanine, Thermo Scientific Chemicals
£31.66 - £273.36
Chemical Identifiers
CAS | 13930-88-6 |
---|---|
Molecular Formula | C32H16N8OV |
Molecular Weight (g/mol) | 579.476 |
MDL Number | MFCD00041966 |
InChI Key | YRZZLAGRKZIJJI-UHFFFAOYSA-N |
Synonym | oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 %Show More |
PubChem CID | 2735160 |
SMILES | C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384497
|
Thermo Scientific Alfa Aesar
039587.02 |
0.5 g |
£31.66
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394497
|
Thermo Scientific Alfa Aesar
039587.04 |
2 g |
£73.16
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374497
|
Thermo Scientific Alfa Aesar
039587.09 |
10 g |
£273.36
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as an organic light-emitting diode intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13930-88-6 | |
579.476 | |
YRZZLAGRKZIJJI-UHFFFAOYSA-N | |
2735160 |
C32H16N8OV | |
MFCD00041966 | |
oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 %Show More | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2]Show More |
Specifications
13930-88-6 | |
Crystalline | |
MFCD00041966 | |
oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 % | |
YRZZLAGRKZIJJI-UHFFFAOYSA-N | |
579.476 | |
579.45 |
Blue to Purple | |
C32H16N8OV | |
UN3285 | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2] | |
2735160 | |
Vanadium(IV) oxide phthalocyanine |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
missing translation for 'einecsNumber' : 237-700-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.