missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 123334-20-3
Molecular Formula O5SV
Molecular Weight (g/mol) 163.00
MDL Number MFCD00149933
InChI Key UUUGYDOQQLOJQA-UHFFFAOYSA-L
Synonym Vanadium oxysulfate hydrate
PubChem CID 16211496
SMILES O=[V++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386098
View Documents
Thermo Scientific Alfa Aesar
011090.09
10 g N/A
N/A
11396098
View Documents
Thermo Scientific Alfa Aesar
011090.18
50 g N/A
N/A
11306108
View Documents
Thermo Scientific Alfa Aesar
011090.30
250 g N/A
N/A
Description

Description

Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123334-20-3
163.00
UUUGYDOQQLOJQA-UHFFFAOYSA-L
16211496
O5SV
MFCD00149933
Vanadium oxysulfate hydrate
O=[V++].[O-]S([O-])(=O)=O

Specifications

123334-20-3
99.9%, metal basis
MFCD00149933
Soluble in water.
O=[V++].[O-]S([O-])(=O)=O
163.00
163.00 (Anhydrous)
Vanadium(IV) Sulfate Oxide Hydrate
Lump
O5SV
Vanadium oxysulfate hydrate
UUUGYDOQQLOJQA-UHFFFAOYSA-L
oxovanadiumbis(ylium) sulfate
16211496
2.5
Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 248-652-7

missing translation for 'rtecsNumber' : YW1840000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Vanadium(IV) sulfate oxide hydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only