missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11386098
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-20-3
Molecular Formula O5SV
Molecular Weight (g/mol) 163.00
MDL Number MFCD00149933
InChI Key UUUGYDOQQLOJQA-UHFFFAOYSA-L
Synonym Vanadium oxysulfate hydrate
PubChem CID 16211496
SMILES O=[V++].[O-]S([O-])(=O)=O

Specifications

Physical Form Lump
Quantity 10 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water.
IUPAC Name oxovanadiumbis(ylium) sulfate
Formula Weight 163.00 (Anhydrous)
Density 2.5
Chemical Name or Material Vanadium(IV) Sulfate Oxide Hydrate
compliance-icons
Product Identifier
  • Vanadium(IV) sulfate oxide hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.