missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(IV,V) oxide, 99.5% (metals basis), Thermo Scientific™

Product Code. 11388227
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 11388227

Brand: Thermo Scientific Alfa Aesar 022956.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-42-2
Molecular Formula O13V6
Molecular Weight (g/mol) 513.64
MDL Number MFCD00064848
InChI Key FTGZPVKRMCXHDZ-UHFFFAOYSA-N
Synonym vanadium oxide v6o13, vanox 13
PubChem CID 72720435
IUPAC Name dioxovanadiooxy(dioxo)vanadium;dioxovanadium
SMILES O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O

Specifications

Melting Point ∼1500°C
Color Black-Blue
Physical Form Crystalline Powder
Quantity 50 g
UN Number UN3285
Solubility Information Soluble in nitric,hydrofluoric,and concentrated sulfuric acids. Insoluble in water.
Formula Weight 513.64
Percent Purity 99.5%
Odor Odorless
Density 3.91 g/mL
Assay (metals basis)
Chemical Name or Material Vanadium(IV,V) oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.