missing translation for 'onlineSavingsMsg'
Learn More
Vanadium(V) oxide, 99.6+%, Thermo Scientific Chemicals Product Code.: 10584464

Vanadium(V) oxide, 99.6+%, Thermo Scientific Chemicals

Product Code. 10584464
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10584464

Brand: Thermo Scientific Acros 206420010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name dioxovanadiooxy(dioxo)vanadium
SMILES O=[V](=O)O[V](=O)=O

Specifications

Boiling Point 1750.0°C
Melting Point 690.0°C
Quantity 1 kg
Assay Percent Range 99.60% min
Linear Formula V2O5
Merck Index 15, 10107
Solubility Information Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies,insoluble in alcohol
Formula Weight 181.88
Percent Purity ≥99.6%
Packaging Glass bottle
Specific Gravity 3.35
Density 3.3500g/mL
Chemical Name or Material Vanadium(V) oxide, 99.6+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468845 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number206422500,206420010
Lot NumberA0465589 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number206422500,206420010
Lot NumberA0464695 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number206422500,206420010
Lot NumberA0413698 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number206422500,206420010
Lot NumberA0406012 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number206422500,206420010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vanadium(V) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity on or via lactation
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Vanadium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(V) oxide, 99.6+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.