Learn More
Vanadium(V) oxytriethoxide, 95%, Thermo Scientific Chemicals
£256.00
Chemical Identifiers
CAS | 1686-22-2 |
---|---|
Molecular Formula | C6H15O4V |
Molecular Weight (g/mol) | 202.12 |
MDL Number | MFCD00799170 |
InChI Key | IDIDIJSLBFQEKY-UHFFFAOYSA-N |
Synonym | triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n |
PubChem CID | 74312 |
IUPAC Name | ethanol;oxovanadium |
SMILES | CCO.CCO.CCO.O=[V] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1686-22-2 | |
202.12 | |
IDIDIJSLBFQEKY-UHFFFAOYSA-N | |
74312 | |
CCO.CCO.CCO.O=[V] |
C6H15O4V | |
MFCD00799170 | |
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n | |
ethanol;oxovanadium |
Specifications
1686-22-2 | |
1.1390g/mL | |
71°C | |
95% | |
(CH3CH2O)3V=O | |
MFCD00799170 | |
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n | |
CCO.CCO.CCO.O=[V] | |
202.12 | |
202.12 | |
Liquid |
Green to Green-Yellow | |
125.0°C to 128.0°C | |
Authentic | |
C6H15O4V | |
1.5080 to 1.5120 | |
1.139 | |
IDIDIJSLBFQEKY-UHFFFAOYSA-N | |
ethanol;oxovanadium | |
74312 | |
95% | |
Vanadium(V) oxytriethoxide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 216-870-1
- Vanadium(V) oxytriethoxide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only