Learn More
Vanadium(V) triisopropoxide oxide, 96%
Description
Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 81°C to 83°C (2mmHg) |
| Melting Point | -12°C |
| Color | Orange-Yellow |
| Physical Form | Liquid |
| Quantity | 25 g |
| Linear Formula | VO[CHO(CH3)2]3 |
| UN Number | UN1993 |
| Flash Point | 45°C (113°F) |
| Solubility Information | Miscible with toluene,hexane and isopropanol. |
| Formula Weight | 242.2 |
| Visa mer |
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.