missing translation for 'onlineSavingsMsg'
Learn More
Vanadium wire, 1.0mm (0.04in) dia, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11326118

Vanadium wire, 1.0mm (0.04in) dia, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11326118
1 m
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy

Product Code. 11326118

Brand: Thermo Scientific Alfa Aesar 011095.G1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]

Specifications

Quantity 1 m
Percent Purity 99.5%
Odor Odorless
Weight ≈4.8g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Vanadium wire, 1.0mm (0.04 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX29K009 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number011095.G5,011095.G1
Lot NumberN22I039 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog Number011095.G5,011095.G1
Lot NumberX16G009 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011095.G5,011095.G1
Lot NumberT30F058 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011095.G5,011095.G1
Lot Number61500275 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011095.G5,011095.G1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium wire, 1.0mm (0.04in) dia, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.