missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl(IV) acetylacetonate, 99%

Product Code. 10338340 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10338340

Brand: Thermo Scientific undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3153-26-2 | C10H14O5V | 265.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-26-2
Molecular Formula C10H14O5V
Molecular Weight (g/mol) 265.16
MDL Number MFCD00000032
InChI Key JFHJZWAQYMGNBE-SUKNRPLKSA-L
Synonym vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide
PubChem CID 131674261
IUPAC Name (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate
SMILES O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 256.0°C to 259.0°C
Color Blue-Green to Green
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Linear Formula (CH3COCH=C(CH3)O)2V=O
Quantity 250 g
Fieser 02,456; 03,331; 11,47; 13,54; 15,57; 15,186
Solubility Information Solubility in water: negligible. Other solubilities: soluble in methanol (6.4 g/100 mL), ethanol, ether, chloroform, acetone, dioxane, benzene (0.9 g/100 m, l), pyridine
Formula Weight 265.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Vanadyl(IV) acetylacetonate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Vanadyl(IV) acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.