missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl(IV) sulfate hydrate, 17-23% V

Product Code. p-40954 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10353382

Brand: Thermo Scientific Acros 214840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12439-96-2 | O5SV·xH2O | 163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12439-96-2
Molecular Formula O5SV·xH2O
Molecular Weight (g/mol) 163
MDL Number MFCD00149933
InChI Key YGYBXHQARYQUAY-UHFFFAOYSA-L
Synonym oxosulfatovanadium pentahydrate, vanadyl sulfate pentahydrate, vanadium, oxosulfato-, pentahydrate, oxovanadium iv sulfate pentahydrate, vanadium 2+ , pentaaquaoxo-, oc-6-21 , sulfate 1:1, oxovanadium iv pentahydrate sulfate, vanadic sulfate pentahydrate, vanadyl sulphate pentahydrate, voso4.5h2o, ov.so4.5h2o
PubChem CID 167150
IUPAC Name oxovanadium(2+);sulfate;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=O)[O-].O=[V+2]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 25 g
Linear Formula VOSO4·xH2O
Solubility Information Solubility in water: soluble
Formula Weight 163
Percent Purity 17 to 23% (V)
Packaging Glass bottle
Chemical Name or Material Vanadyl(IV) sulfate hydrate, 17-23% V
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vanadyl(IV) sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.