missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg Product Code.: 10379350

Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg

Product Code. 10379350
1 g, Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10379350

Brand: Thermo Scientific Chemicals 296990010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vancomycin hydrochloride (>900 microgram/mg), CAS # 1404-93-9, is an antibiotic from Streptomyces orientalis, that shows antibacterial activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic and antiviral from Streptomyces orientalis
  • It shows antibiotic activity towards methicillin-resistant (β-lactam-resistant) Staphylococci bacterial strains
  • Vancomycin hydrochloride inhibits bacterial mucopeptide synthesis

Applications

  • Vancomycin hydrochloride demonstrates potent activity against gram-positive bacteria
  • It has been shown in studies to be highly effective against methicillin-resistant Staphylococcus aureus (MRSA) in vitro and in vivo
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
pH 2.5 to 4.5 (5% soln.)
Infrared Spectrum Authentic
Packaging Glass Bottle
Water 5% max.
Color Tan to White
Quantity 1 g
Solubility Information Solubility in water: 50g/L. Other solubilities: moderately soluble in diluted methanol, insoluble in higher alcohols, acetone and ether
Formula Weight 1485.73
Percent Purity ≥80.0% (vancomycin B) (HPLC), 9.0% (individual impurities) max., greater than 900μg/mg (potency, on anhydrous substance)
Chemical Name or Material Vancomycin hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467816 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number296990010,296990050
Lot NumberA0462934 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number296990010,296990050
Lot NumberA0446191 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number296990010,296990050
Lot NumberA0399356 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number296990010,296990050
Lot NumberA0396051 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number296990010,296990050
compliance-icons
Product Identifier
  • Vancomycin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Vancomycin hydrochloride, >900 microgram/mg >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.