missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Vandetanib Product Code.: 16594404

Thermo Scientific Chemicals Vandetanib

Product Code. 16594404
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 16594404

Brand: Thermo Scientific Chemicals 464332500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443913-73-3
Molecular Formula C22H24BrFN4O2
Molecular Weight (g/mol) 475.36
InChI Key UHTHHESEBZOYNR-UHFFFAOYSA-N
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
SMILES COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1

Specifications

Quantity 250 mg
Melting Point 240.0°C to 243.0°C
Solubility Soluble in methanol
Formula Weight 475.35
Chemical Name or Material Vandetanib

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0408668 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog Number464332500,464330010,464330050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Vandetanib >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.