missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testo™ Vane Anemometer
Brand: Testo™ 0560 4053
127.29 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 04
Additional Details : Weight : 0.20000kg
Specifications
Thermal Anemometer | |
490 x 37 x 36 mm | |
-20 to +70°C |
±0.5 m/sec. | |
Measurement of indoor air velocity, Measurement in ventilation ducts |