missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Inlet Check Valves for Solvent Management
VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling
Specifications
Type | Inlet Check Valve; 0.1-0.25 psi crack pressure |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Solvent Containment Systems |
Includes | 1 Ea. |
Thread Size (English) | 1/4 in.-28 UNF(M) |
Description
- Minimize evaporation and relieve pressure to improve safety
- Allow air into supply container to displace vacuum and keep solvent flowing
Warranty
None
Specifications
Inlet Check Valve; 0.1-0.25 psi crack pressure | |
VapLock Solvent Containment Systems | |
1/4 in.-28 UNF(M) |
Solvent Safety | |
1 Ea. |