missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Relief Valves for Solvent Management

VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling

£73.30

Specifications

Type Relief Valve
For Use With (Application) Solvent Safety
For Use With (Equipment) VapLock Solvent Containment Systems
Thread Size (English) 1/4 in.-28 UNF(M)
View More Specs

Products 1
Product Code Brand Material Pressure Price Quantity & Availability  
Product Code Brand Material Pressure Price Quantity & Availability  
15894434
View Documents
Cole-Parmer™
12020-62
PTFE, FFKM 0.1 to 0.25 psi crack pressure
£73.30
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
 
Description

Description

Choice of 0.1 to 0.25 psi or 1 to 2.5 psi cracking pressure

Warranty

None

Specifications

Specifications

Relief Valve
Solvent Safety
VapLock Solvent Containment Systems
1/4 in.-28 UNF(M)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.