missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Reversible Valves for Solvent Management
VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling
Specifications
Type | Reversible Valve |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Solvent Containment Systems |
Pressure | 0.1 to 0.25 psi crack pressure |
Thread Size (English) | 2 x 1/4 in. NPT(M) x 1/4 in. NPT(M) |
Description
- Minimize evaporation and relieve pressure to improve safety
- Change orientation to switch between inlet and outlet flow regulation
Warranty
None
Specifications
Reversible Valve | |
VapLock Solvent Containment Systems | |
2 x 1/4 in. NPT(M) x 1/4 in. NPT(M) |
Solvent Safety | |
0.1 to 0.25 psi crack pressure |