missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps
VapLock™ Solvent Delivery Caps reduce chemical evaporation and exposure to hazardous vapors
Brand: Cole-Parmer™ 12018-01
74.30 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.11000kg
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODServices
None
Specifications
Solvent Safety | |
304 SS Port Thread Inserts, three 1/4 in.-28 | |
Solvent-Compatible Containers |
GL45 | |
Blue/white PTFE, PP, SS | |
1/4 in.-28 UNF(F) |