missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Drums

VapLock™ Solvent Waste Manifold Caps for Drums connect to waste containers for a secure closed system

Specifications

Type Solvent Waste Manifold
For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
View More Specs

Description

Description

  • Select from coarse or fine thread to fit your drum's 2 in bung
  • Different materials, numbers and sizes of ports help customize to your application

Warranty

None

Specifications

Specifications

Solvent Waste Manifold
Solvent Safety
Solvent-Compatible Containers
Videos
SDS
Documents

Documents

Product Certifications