missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ VapLock™ Threaded Plugs

Use VapLock™ Threaded Plugs to seal unused ports for a leak-resistant system

£84.62

Specifications

Type Port Plug
For Use With (Application) Solvent Safety
For Use With (Equipment) VapLock Solvent Containment Systems
View More Specs

Products 1
Product Code Brand Material Thread Size (English) Price Quantity & Availability  
Product Code Brand Material Thread Size (English) Price Quantity & Availability  
15864464
View Documents
Cole-Parmer™
12020-47
PTFE 1/4 in.-28 UNF(M)
£84.62
Pack of 10
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
 
Description

Description

Seal unused manifold and bottle cap ports

Warranty

None

Specifications

Specifications

Port Plug
Solvent Safety
VapLock Solvent Containment Systems
Videos
SDS
Documents

Documents

Product Certifications