missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varenicline tartrate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Orally active, subtype-selective partial agonist at α4β2 nicotinic receptors (Ki values are 0.06, 240, 322 and 3540 nM for α4β2, α3β4, α7, α1βγδ receptors respectively). Reduces nicotine-evoked dopamine release in vitro and decreases nicotine self-administration in vivo. Also potently activates PSAM4-5-HT3 and PSAM4-GlyR chimeric ion channels (EC50 values are 4 and 1.6 nM, respectively)
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 361.37 |
Percent Purity | >99% |
Chemical Name or Material | Varenicline tartrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'