missing translation for 'onlineSavingsMsg'
Learn More

Varenicline tartrate, Tocris Bioscience™

Product Code. 18752504
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective α4β2 partial agonist; orally active

Orally active, subtype-selective partial agonist at α4β2 nicotinic receptors (Ki values are 0.06, 240, 322 and 3540 nM for α4β2, α3β4, α7, α1βγδ receptors respectively). Reduces nicotine-evoked dopamine release in vitro and decreases nicotine self-administration in vivo. Also potently activates PSAM4-5-HT3 and PSAM4-GlyR chimeric ion channels (EC50 values are 4 and 1.6 nM, respectively)

Chemical Identifiers

CAS 375815-87-5
Molecular Formula C17H19N3O6
Molecular Weight (g/mol) 361.354
InChI Key TWYFGYXQSYOKLK-ZBABXEQTSA-N
Synonym varenicline tartrate
PubChem CID 124080896
SMILES C1C2CNCC1C3=CC4=NC=CN=C4C=C23.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Quantity 50 mg
Formula Weight 361.37
Percent Purity >99%
Chemical Name or Material Varenicline tartrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'