Learn More
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings
For the disintegration of COD, total nitrogen and total phosphorus
Brand:  WTW™ 1P21-1
 View more versions of this product
Product Code. 10159081
  • £1025.00 / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermoreactors for the disintegration of COD, total nitrogen and total phosphorus, including brief and self-programmed high temperature disintegration up to 170°C. The high reaction temperature over a defined period of time ensures a complete degradation of the sample. The required temperatures and degradation times for the standard parameters are stored in every WTW thermoreactor. In addition, there are different options for self programming and cuvette numbers available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermoreactor
230/115 V
315 mm
Routine measurements, wastewater
12 Reaction Cuvettes
50/60 Hz
280 W
115/230 V ±15%, 50/60 Hz, 280 W
185 mm
256 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products