missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Varian Split Liner with Quartz Wool
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™ 09202101
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Varian instruments
Specifications
GC Inlet Liner, Split Liner with Quartz Wool | |
72 mm | |
6.3 mm |
1 Ea. | |
4 mm | |
Varian 1075/1077 |