Learn More
Macherey-Nagel™ VarioPrep™ Guard Column Holder for Preperative HPLC
Use VarioPrep™ stainless steel columns for HPLC. VarioPrep™ Guard Column Holder for Preperative HPLC come in different sizes for a variety of columns.
£234.00 - £401.00
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
12757690
|
Macherey-Nagel™
718253 |
32 mm | 15 mm |
£401.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
12472610
|
Macherey-Nagel™
718251 |
8 mm | 10 mm |
£234.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
||||
Description
Features- Column system for preparative HPLC manufactured from stainless steel with two adjustable end fittings, e.g., for frequent use of backflushing techniques
- Allows compensation of a dead volume, which could result at the column inlet after some time of operation, without need for opening the column
- Can be packed with all NUCLEODUR™ and NUCLEOSIL™ spherical silicas
- Column holders come in a variety of sizes for different columns
Specifications
Column Holder |