Learn More
Eppendorf™ Varitip™ S Starter Kit
Tailored to different vessels. Choose from a variety of Eppendorf Varitips to meet your needs.
Brand: Eppendorf™ 0030050525
This item is not returnable.
View return policy
Description
- Eppendorf Varitip P is designed for aspirating 1 mL to 10 mL from beakers
- Eppendorf Varitip S for 2.5 mL to 10 mL forms a system with the Varitip and is used for aspirating liquid from tall, narrow-necked vessels
Specifications
Eppendorf Varitips S Starter Kit | |
Removing liquid from narrow-necked vessels and measuring flasks, | |
100 Maxitips, 10 dispensing parts, 10 valves | |
1 Kit |