missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Vaseline for Rotors

Product Code. 30130221 Shop All Ohaus Products
Click to view available options
Quantity:
10 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30130221

Brand: OHAUS™ 30314586

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High-quality product designed to enhance the performance and durability of your rotors. This specially formulated Vaseline is the perfect solution for maintaining the smooth operation of your sophisticated laboratory equipment.

This Vaseline for rotors is a high-quality product designed to enhance the performance and durability of your rotors. This specially formulated Vaseline is the perfect solution for maintaining the smooth operation of your sophisticated laboratory equipment.

  • Crafted with meticulous attention to detail, this Vaseline is an essential tool for any researcher, scientist, or technician. It is designed to provide excellent lubrication, reducing friction and wear on the rotor's moving components. By protecting against corrosion and ensuring optimal operation, this product guarantees long-lasting performance for your valuable equipment.
  • One of the outstanding features is its exceptional versatility. Suitable for use with a wide range of rotors, it is a highly reliable choice for various laboratory applications. Whether you are working with centrifuges, emulsifiers, or any other complex rotor machinery, this Vaseline will deliver exceptional results.
  • In addition to its exceptional lubricating properties, this Vaseline boasts superior chemical resistance and stability. It can withstand extreme temperatures, ensuring consistent performance in demanding laboratory environments. The high-quality formulation prevents the Vaseline from breaking down or losing its effectiveness, even in the harshest conditions.
  • Comes in a convenient, easy-to-use container that makes application a breeze. The precise nozzle allows you to control the amount of Vaseline applied, preventing wastage and ensuring optimal coverage. With its long shelf life and manageable container size, this Vaseline is the perfect companion for your laboratory, giving you peace of mind while conducting your experiments.
  • Investing in this Vaseline means investing in the longevity and efficiency of your laboratory equipment. Trust the world-renowned OHAUS brand to deliver a top-quality product that meets the highest standards of performance, reliability, and safety. With this Vaseline, you can focus on your research without worrying about equipment malfunctions or unnecessary downtime.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lubricant
Format Viscous Liquid
For Use With (Application) Lubricating Rotors
For Use With (Equipment) Rotors
Quantity 10 mL
Volume (English) 10 mL
Volume (Metric) 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.