missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ VCL/ALK DY Translocation FISH Probe
Labeled FISH probes for identification of gene transloaction using Fluoresecent In Situ Hybridization Technique.
£1285.00 - £2110.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 7414/238 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083565
|
Abnova™
FT0019.100uL |
100 μL |
£1285.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093565
|
Abnova™
FT0019.200uL |
200 μL |
£2110.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark. | |
Tube | |
7414/238 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |