Learn More
Velp Scientifica™ Vcopper
Velp Scientifica™ Vcopper for use with Dumas Nitrogen Analyzer NDA 701 and 702.
Brand: Velp Scientifica™
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.47000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
- For use with NDA 701 and 702
Specifications
Vcopper | |
470 g | |
Dumas Nitrogen Analyzer | |
0.47 kg |
Safety and Handling
- Vcopper High Reduction Efficiency
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper