missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ VCP 171
Positive allosteric modulator (PAM) of A1 receptors; also allosteric partial agonist in absence of orthosteric agonist
£156.58 - £624.17
Specifications
Chemical Name or Material | [2-Amino-4-[3-(trifluoromethyl)phenyl]-3-thienyl]phenylmethanone |
---|---|
CAS | 1018830-99-3 |
Target | Adenosine A1 Receptor Modulators |
Molecular Formula | C18H12F3NOS |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779916
|
Tocris™
6261/10 |
10 mg |
£156.58
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759886
|
Tocris™
6261/50 |
50 mg |
£624.17
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VCP 171 is an adenosine A1 receptor positive allosteric modulator (PAM) (pKB = 5.65, binding cooperativity with NECA 0.68). In absence of orthosteric agonist, VCP171 behaves as an allosteric partial agonist, inhibiting cAMP activity. Inhibits AMPA receptor-mediated evoked excitatory postsynaptic current (eEPSC) amplitude in lamina I and II neurons in a rat partial nerve-injury model of neuropathic pain.Specifications
[2-Amino-4-[3-(trifluoromethyl)phenyl]-3-thienyl]phenylmethanone | |
Adenosine A1 Receptor Modulators | |
0.98 |
1018830-99-3 | |
C18H12F3NOS |
Safety and Handling
Recommended Storage : Store at -20°C