Learn More
VD3-d6, MedChemExpress
MedChemExpress VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3; tools for determination of Vitamin D3 metabolites in human serum.
POA - £2703.14
Specifications
Chemical Name or Material | VD3-d6 |
---|---|
CAS | 118584-54-6 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C27H38D6O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16469047
|
Medchem Express
HY-15331 |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16431509
|
Medchem Express
HY-15331 |
1 mg |
£2703.14
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VD3-d6 (Vitamin D3-26,26,26,27,27,27-d6) is the deuterium labeled Vitamin D3Specifications
VD3-d6 | |
Light Yellow | |
C27H38D6O | |
DMSO : 100 mg/mL (255.97 mM; Need ultrasonic) | |
390.67 | |
98.0% | |
Neuroscience-Neuromodulation |
118584-54-6 | |
Research | |
Vitamin D3-26,26,26,27,27,27-d6 | |
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2 | |
390.67 | |
Research | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.