missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Receptor 3 Monoclonal Antibody (44CT92.4.4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511168
This item is not returnable.
View return policy
Description
Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.Specifications
VEGF Receptor 3 | |
Monoclonal | |
0.44 mg/mL | |
PBS with 0.09% sodium azide | |
P35916 | |
FLT4 | |
Purified His-tagged VGFR3 protein fragment | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
44CT92.4.4 | |
Unconjugated | |
FLT4 | |
AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3 | |
Mouse | |
Protein G | |
RUO | |
2324 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |