Learn More
Velp Scientifica™ OXISoft™ Software
Velp Scientifica™ OXISoft™ Software manages all analysis steps and readily provides valuable information to scientists, for use with OHS Controllers
Brand:  Velp Scientifica™ A00000391
Product Code. 16665442
  • £264.00 / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This software manages all analysis steps and readily provides valuable information to scientists conducting experiments
  • Enables scientists to perform repeatability tests, freshness tests, formula comparison, packaging comparison, IP during ageing, and estimated shelf life
  • Up to four tests can be managed by the same software for maximum productivity
Specifications
Specifications
OHS Controllers
Software