missing translation for 'onlineSavingsMsg'
Learn More

Velsecorat, MedChemExpress

MedChemExpress AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM.

Encompass_Preferred

£192.39 - £1501.61

Specifications

Chemical Name or Material Velsecorat
CAS 1196509-60-0
Color White
Purity Grade Notes Research
Molecular Formula C32H32F2N4O6
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16497177
Encompass_Preferred
View Documents
Medchem Express
HY-111453
5 mg
£192.39
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16495336
Encompass_Preferred
View Documents
Medchem Express
HY-111453
100 mg
£1501.61
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16454458
Encompass_Preferred
View Documents
Medchem Express
HY-111453
10 mg
£263.78
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16433096
Encompass_Preferred
View Documents
Medchem Express
HY-111453
25 mg
£589.27
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16400777
Encompass_Preferred
View Documents
Medchem Express
HY-111453
50 mg
£948.64
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AZD7594 is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM.
Specifications

Specifications

Velsecorat
White
C32H32F2N4O6
DMSO : ≥ 83.3 mg/mL (137.32 mM)
606.62
99.6%
Solid
1196509-60-0
Research
AZD7594 AZ13189620
O=C(N[C@H]1COCC1)C2=CC=CC(N3N=CC4=C3C=CC(O[C@H](C5=CC=C(OCCO6)C6=C5)[C@@H](NC(C(F)(F)C)=O)C)=C4)=C2
606.62
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Velsecorat, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.